Learn More
E-982, MedChemExpress
Description
E-982 is a steroid used for the on-line screening of the DNA unwinding element binding protein (DUE-B) immobilized protein column.
Specifications
Specifications
| Chemical Name or Material | E-982 |
| CAS | 858102-78-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H31NO6S |
| Quantity | 5 mg |
| SMILES | OC(C=C1)=CC2=C1[C@@]3([H])CC[C@]4(C)[C@@H](O)CC[C@@]4([H])[C@]3([H])C(SCCC(ON5C(CCC5=O)=O)=O)C2 |
| Molecular Weight (g/mol) | 473.58 |
| Formula Weight | 473.58 |
| Percent Purity | 98.0% |
| Show More |
Research purposes only