Learn More
3M™ E-A-R™ Tri-Flange™ Earplugs
Click to view available options
Cord Material:
Cloth
Vinyl
Unit Size:
100 pairs
Description
- Earplugs with triple flanges that adapt to the shape of the ear to form multiple seals against noise
- No need to handle the plug tip making them more hygienically acceptable
- Triple-flange design preformed to fit the ear
- Reusable plug is comfortable, hygienic and economical as it can be washed
- Supplied with a cotton or vinyl cord
Specifications
Specifications
| Color | Yellow |
| Certifications/Compliance | CE Approved |
| Cord Material | Cloth |
| Pairs per Package | 400 Pairs/Box |
| Noise Reduction Rating (NRR) | 29 dB |
| Dielectric | No |
| Material | Elastomeric Polymer |