missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin Antibody (CDH1/4398R), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-09082JF646
This item is not returnable.
View return policy
Description
E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| E-Cadherin | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CDH1/4398R | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
| Synthetic peptide corresponding to E-Cadherin residues within aa600-700 of E-cadherin (Uniprot: P12830) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 999 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction