missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin Antibody (CL1180), Novus Biologicals™
Mouse Monoclonal Antibody
€ 285.00 - € 413.00
Specifications
| Antigen | E-Cadherin |
|---|---|
| Clone | CL1180 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18424662
|
Novus Biologicals
NBP2-34477-25ul |
25 μL |
€ 285.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18174162
|
Novus Biologicals
NBP2-34477 |
0.1 mL |
€ 413.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| E-Cadherin | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| P12830 | |
| 999 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ATDNGSPVATGTGTLLLILSDVNDNAPIPEPRTIFFCERNPKPQVINIIDADLPPNTSPFTAELTHGASANWTIQYNDPTQESIILKPKMALEVGDYKINLKLMDNQNKDQVTTLEVSVCDCEGA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL1180 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
| CDH1 | |
| IgG2a | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title