Learn More
(E)-Cefpodoxime Proxetil, TRC
Product Code. 30365042
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365042 1 mg 1mg
30360894 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30365042 Supplier Toronto Research Chemicals Supplier No. TRCC2438551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 947692-13-9
Molecular Formula C21H27N5O9S2
Molecular Weight (g/mol) 557.6
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2E)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-, 1-[[(1-methylethoxy)carbonyl]oxy]ethyl ester, (6R,7R)-, 1-[[(1-Methylethoxy)carbonyl]oxy]ethyl (6R,7R)-7-[[(2E)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, (1RS)-1-[[(1-Methylethoxy)carbonyl]oxy]ethyl (6R,7R)-7-[[(2E)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, anti-Cefpodoxime Proxetil, (E)-Cefpodoxime Proxetil, Cefpodoxime Proxetil Imp. D (EP)
IUPAC Name 1-isopropoxycarbonyloxyethyl (6R,7R)-7-[[(2E)-2-(2-aminothiazol-4-yl)-2-methoxyimino-acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES COCC1=C(N2[C@H](SC1)[C@H](NC(=O)\C(=N\OC)\c3csc(N)n3)C2=O)C(=O)OC(C)OC(=O)OC(C)C
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C21H27N5O9S2/c1-9(2)33-21(30)35-10(3)34-19(29)15-11(6-31-4)7-36-18-14(17(28)26(15)18)24-16(27)13(25-32-5)12-8-37-20(22)23-12/h8-10,14,18H,6-7H2,1-5H3,(H2,22,23)(H,24,27)/b25-13+/t10?,14-,18-/m1/s1
Formula Weight 557.125
Chemical Name or Material (E)-Cefpodoxime Proxetil