missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E. Coli K88A Antibody (1050/66), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64537JF549
This item is not returnable.
View return policy
Description
E. Coli K88A Monoclonal specifically detects E. Coli K88A in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| E.ColiK88A | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| faeC | |
| NCTC 10964 (Nobivac) | |
| 0.1 mL | |
| In a simple ELISA, using Biogenesis test groups, this is only reactive with E. coli K88a. Reactive with isolates fixed by phenol, formalin and boiling. | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 1050/66 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Escherichia coli | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction