missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E. Coli Labile Toxin A Chain Antibody (20/480) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64577
This item is not returnable.
View return policy
Description
E. Coli Labile Toxin A Chain Monoclonal specifically detects E. Coli Labile Toxin A Chain in Bacteria samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| E. Coli Labile Toxin A Chain | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Escherichia coli | |
| Mouse | |
| Protein A purified | |
| RUO | |
| In a simple ELISA, using Biogenesis test groups, this is only reactive with E. coli labile toxin, C terminal of A chain. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 20/480 | |
| Unconjugated | |
| P43530 | |
| eltA | |
| Native, labile toxin | |
| 0.2 mg | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction