missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E. coli (strain K12/DH10B) NfuA Polyclonal Antibody
GREENER_CHOICE

Product Code. 17210154 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17210154 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17210154 Supplier Invitrogen™ Supplier No. PA5117652

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Involved in iron-sulfur cluster biogenesis. Binds a 4Fe-4S cluster, can transfer this cluster to apoproteins, and thereby intervenes in the maturation of FeS proteins. Could also act as a scaffoldchaperone for damaged FeS proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli (strain K12/DH10B) NfuA
Applications Western Blot
Classification Polyclonal
Concentration 2.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene nfuA
Gene Accession No. B1X760
Gene Alias Fe/S biogenesis protein NfuA nfuA ECDH10B_3589
Gene Symbols nfuA
Host Species Rabbit
Immunogen Recombinant Escherichia coli Fe/S biogenesis protein NfuA protein (1-191aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58390684, 82263228
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.