Learn More
(E)-Diethyl (2-Butenyl)phosphonate, TRC
Specifications
Specifications
| Chemical Name or Material | (E)-Diethyl (2-Butenyl)phosphonate |
| CAS | 26327-86-6 |
| Molecular Formula | C8H17O3P |
| Quantity | 5 g |
| Synonym | Diethyl trans-2-butenylphosphonate, (E)-2-Butenyl-phosphonic Acid Diethyl Ester, (2E)-2-Butenyl-phosphonic Acid Diethyl Ester |
| InChI Formula | InChI=1S/C8H17O3P/c1-4-7-8-12(9,10-5-2)11-6-3/h4,7H,5-6,8H2,1-3H3/b7-4+ |
| SMILES | CCOP(C/C=C/C)(OCC)=O |
| IUPAC Name | diethyl (E)-but-2-en-1-ylphosphonate |
| Molecular Weight (g/mol) | 192.19 |
| Formula Weight | 192.09 |