Learn More
Invitrogen™ E-Gel™ High-Throughput Mix Pack
Beschreibung
The E-Gel High-Throughput Mix Pack allows researchers to try all of our high-throughput format E-Gel agarose gels. These gels use SYBR safe as a DNA stain to minimize exposure to hazardous and mutagenic factors such as ethidium bromide or UV light. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.
Benefits of using E-Gel agarose gels with SYBR Safe include:
- Better sample integrity—blue-light excitation wavelength minimizes DNA damage, as opposed to UV light
- Safer choice—contains SYBR Safe DNA stain, a safer alternative to ethidium bromide
- High performance—detect 3 ng of sample DNA with a separation time as short as 12 minutes
- No buffers—E-Gel agarose gels do not require buffers to run
The box contains:
- E-Gel 48 Agarose Gels with SYBR Safe DNA Gel Stain, 1%
- E-Gel 48 Agarose Gels with SYBR Safe DNA Gel Stain, 2%
- E-Gel 96 Agarose Gels with SYBR Safe DNA Gel Stain, 1%
- E-Gel 96 Agarose Gels with SYBR Safe DNA Gel Stain, 2%
Running and imaging devices
High-throughput E-Gel agarose gels with SYBR Safe require the E-Gel Power Snap Plus Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Spezifikation
Spezifikation
| Format | Kit |
| Gel Type | E-Gel |
| Separation of | DNA |
| Separation Range | 100 to 2000 bp |
| Separation Type | Electrophoresis |
| Quantity | 8 Gels |
| Sterility | Non-sterile |