Learn More
Bochem™ E-Poli Test Tube Stands
Click to view available options
Array:
10 x 10
2 x 10
2 x 12
3 x 12
4 x 12
4 x 6
Unit Size:
Each
Specifications
Specifications
| Holds | Tubes (16 mm) |
| Material | 18/10 E-Poli |
| Positions | 100 |
| Diameter (Metric) Opening | 16 mm |
| Product Type | Test Tube Rack |
| Array | 10 x 10 |