missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Selectin/CD62E Antibody (105), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90951AF532
This item is not returnable.
View return policy
Description
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Cynomolgus Monkey samples. It is validated for ELISA.
Specifications
| E-Selectin/CD62E | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus E-Selectin/CD62E (G8F370; Met1-Pro556). | |
| 0.1 mL | |
| Immunology, Lipid and Metabolism | |
| 6401 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 105 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction