missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Selectin/CD62E Antibody (208), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89432PE
This item is not returnable.
View return policy
Description
E-Selectin/CD62E Monoclonal antibody specifically detects E-Selectin/CD62E in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| E-Selectin/CD62E | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 208 | |
| Immunocytochemistry/ Immunofluorescence | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human E-Selectin/CD62E (Accession#: NP_000441.2; Met1-Pro556). | |
| 0.1 mL | |
| Immunology, Lipid and Metabolism | |
| 6401 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction