missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Selectin/CD62E Antibody (RM0034-9J12), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-85473MFV450
This item is not returnable.
View return policy
Description
E-Selectin/CD62E Monoclonal antibody specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Flow Cytometry
Specifications
| E-Selectin/CD62E | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| RM0034-9J12 | |
| Flow Cytometry | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
| Cytokine-stimulated mouse E-Selectin/CD62E | |
| 0.1 mL | |
| Immunology, Lipid and Metabolism | |
| 6401 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction