missing translation for 'onlineSavingsMsg'
Learn More

(E)-Toremifene, (E)-Toremifene, TRC

Product Code. 30137150
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137150 10 mg 10mg
30006182 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137150 Supplier Toronto Research Chemicals Supplier No. TRCT54749010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C26H28ClNO
Molecular Weight (g/mol) 405.96
Synonym 2-[4-[(1E)-4-Chloro-1,2-diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethylethanamine;
IUPAC Name 2-[4-[(E)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine
SMILES CN(C)CCOc1ccc(cc1)\C(=C(/CCCl)\c2ccccc2)\c3ccccc3

Specifications

CAS 89778-29-0
Quantity 10 mg
InChI Formula InChI=1S/C26H28ClNO/c1-28(2)19-20-29-24-15-13-23(14-16-24)26(22-11-7-4-8-12-22)25(17-18-27)21-9-5-3-6-10-21/h3-16H,17-20H2,1-2H3/b26-25+
Formula Weight 405.1859
Chemical Name or Material (E)-Toremifene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.