Learn More
(E,Z)-Methomyl-d3 (N-methyl-d3), CDN
Product Code. 30326783
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326783 0.01 g 0.01g
30313898 0.05 g 0.05g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30326783 Supplier CDN Supplier No. CDND74540.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methomyl D3
CAS 1398109-07-3
Molecular Formula C5 2H3 H7 N2 O2 S
Quantity 0.01 g
Synonym Methomyl D3
InChI Formula InChI=1 S/C5H10N2O2S/c1-4(10-3)7-9-5(8)6-2/h1-3H3,(H,6,8)/i2D3
SMILES [2 H]C([2 H])([2 H])NC(=O)ON=C(C)SC
IUPAC Name methyl N-(trideuteriomethylcarbamoyloxy)ethanimidothioate
Molecular Weight (g/mol) 165.23
Formula Weight 165.0651 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity