missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F-4 Antibody (E2F4/8811R) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26926JF525
This item is not returnable.
View return policy
Description
E2F-4 Monoclonal antibody specifically detects E2F-4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| E2F-4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| E2F transcription factor 4, p107/p130-binding, E2F-4p107/p130-binding protein, transcription factor E2F4 | |
| Recombinant fragment (around aa1-100) of human E2F-4 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1874 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| E2F4/8811R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction