Learn More
E2F1 Recombinant Rabbit Monoclonal Antibody (23GB1505), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen MA553964
 View more versions of this product
Product Code. 30088655
  • € 562.00 / 100µL
Description
E2F1 Recombinant Monoclonal antibody specifically detects E2F1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E2F1
Recombinant Monoclonal
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O09139, Q01094, Q61501
E2F1
A synthesized peptide derived from human E2F1 (355-400AA).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB1505
Unconjugated
E2F1
E2F transcription factor 1; E2F1; E2F-1; mKIAA4009; PBR3; PRB-binding protein E2F-1; RBAP1; RBAP-1; RBBP3; RBBP-3; RBP3; Retinoblastoma-associated protein 1; retinoblastoma-binding protein 3; Transcription factor E2F1
Rabbit
Affinity Chromatography
RUO
13555, 1869, 399489
-20°C