missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-12 p80 Recombinant Protein, PeproTech®

Product Code. 30088085
Click to view available options
Quantity:
10 x 50 μg
2 x 50 μg
5 x 50 μg
Unit Size:
Pack of 10
Pack of 2
Pack of 5
This item is not returnable. View return policy

Product Code. 30088085

Brand: Gibco™ 21012P80H500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Accumulating data indicates that p40 secretion precedes that of IL-12 expression. In addition, to its ability to covalently bind to p35 to form IL-12, p40 can bind to p19 to form IL-23, or it can form the homodimer designated IL-12 p80. Elevated levels of IL-12 p80 correlate to macrophage recruitment and increased inflammation in asthma and respiratory viral infection models.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43432
For Use With (Application) Functional Assay
Formulation Protein with no preservative
Gene ID (Entrez) 16160
Molecular Weight (g/mol) 80 kDa
Name IL-12 p80
Quantity 10 x 50 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias IL-12; Interleukin
Gene Symbol IL12B
Conjugate Unconjugated
Recombinant Recombinant
Activity Determined by its ability to inhibit the proliferative effect of 2 ng/ml mIL-12 p70 in T cell-enriched PBMCs.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.