missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111387
This item is not returnable.
View return policy
Description
Immunogen sequence: GPNPSTSFEP IKADPTGVLE LPKELSEIFD PTRECMSSEL LEELMSSEVF APLLRLSPPP GDHDYIYNLD ESEGVCD Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).
Specifications
| E2F4 | |
| Polyclonal | |
| Unconjugated | |
| E2f4 | |
| 2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1874 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16254 | |
| E2f4 | |
| Recombinant Protein corresonding to Human E2F4. Recombinant protein control fragment (Product #RP-106509). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction