missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143103
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).
Specifications
| E2F4 | |
| Polyclonal | |
| Unconjugated | |
| E2f4 | |
| 2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 100522677, 1874 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q16254 | |
| E2f4 | |
| Peptide with sequence C-PKELSEIFDPTR. | |
| 100 μg | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction