missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10923-1-AP-20UL
This item is not returnable.
View return policy
Description
E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).Specifications
| E2F4 | |
| Polyclonal | |
| Unconjugated | |
| E2F4 | |
| E2F 4, E2F4, Transcription factor E2F4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100360427, 104394, 1874 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16254, Q8R0K9 | |
| E2F4 | |
| E2F4 Fusion Protein Ag1365 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction