missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F4 Recombinant Rabbit Monoclonal Antibody (3O6Y9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542723
This item is not returnable.
View return policy
Description
Immunogen sequence: TAVPGSAEVQ GMAGPAAEIT VSGGPGTDSK DSGELSSLPL GPTTLDTRPL QSSALLDSSS SSSSSSSSSS NSNSSSSSGP NPSTSFEPIK ADPTGVLELP K.
E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).
Specifications
| E2F4 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16254, Q8R0K9 | |
| E2f4 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3O6Y9 | |
| Unconjugated | |
| E2f4 | |
| 2010111M04Rik; AI427446; E2F transcription factor 4; E2F transcription factor 4, p107/p130-binding; E2F4; E2F-4; p107/p130-binding protein; transcription factor E2F4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 104394, 1874 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction