missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA581166
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human E2F5.
E2F5 Polyclonal antibody specifically detects E2F5 in Human samples. It is validated for Immunoprecipitation, Western Blot.
Specifications
| E2F5 | |
| Polyclonal | |
| Unconjugated | |
| E2F5 | |
| AU024671; E2F transcription factor 5; E2F transcription factor 5, p130-binding; E2f5; E2F-5; Transcription factor E2F5 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1875 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 5 mg/mL | |
| PBS with no preservative | |
| Q15329 | |
| E2F5 | |
| A synthetic peptide corresponding to the N-terminus of the Human E2F5 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction