missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F6 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99087CL1
This item is not returnable.
View return policy
Description
E2F6 Polyclonal antibody specifically detects E2F6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| E2F6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| E2F transcription factor 6, E2F-6MGC111545, transcription factor E2F6 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human E2F6. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1876 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction