missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F6 Antibody (PCRP-E2F6-1F8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08952B
This item is not returnable.
View return policy
Description
E2F6 Monoclonal antibody specifically detects E2F6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| E2F6 | |
| Monoclonal | |
| Biotin | |
| E2F transcription factor 6, E2F-6MGC111545, transcription factor E2F6 | |
| Recombinant full-length human E2F6 protein (Uniprot: O75461) | |
| 100 μg | |
| Cell Cycle and Replication | |
| 1876 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-E2F6-1F8 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction