missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E2F6 Recombinant Rabbit Monoclonal Antibody (9I5M0)
GREENER_CHOICE

Product Code. 17286713 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17286713 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17286713 Supplier Invitrogen™ Supplier No. MA537840

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Samples: 293T, HepG2, RD Immunogen sequence: LDGIDLVEKK SKNHIRWIGS DLSNFGAVPQ QKKLQEELSD LSAMEDALDE LIKDCAQQLF ELTDDKENER LAYVTYQDIH SIQAFHEQIV IAVKAPAETR LDVPAPREDS ITVHIRSTNG PIDVYLCEVE QGQTSNKRSE GVGTSSSEST HPEGPEEEEN PQQSEELLEV SN.

This gene encodes a member of the E2F transcription factor protein family. E2F family members play a crucial role in control of the cell cycle and of the action of tumor suppressor proteins. They are also a target of the transforming proteins of small DNA tumor viruses. Many E2F proteins contain several evolutionarily conserved domains: a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. The encoded protein of this gene is atypical because it lacks the transactivation and tumor suppressor protein association domains. It contains a modular suppression domain and is an inhibitor of E2F-dependent transcription. The protein is part of a multimeric protein complex that contains a histone methyltransferase and the transcription factors Mga and Max. Multiple transcript variants have been reported for this gene, but it has not been clearly demonstrated that they encode valid isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F6
Applications ChIP Assay, ELISA, Western Blot, CUT&RUN
Classification Recombinant Monoclonal
Clone 9I5M0
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene E2F6
Gene Accession No. O75461
Gene Alias AI462434; E2F transcription factor 6; E2f6; E2F-6; E2F6b; E2F-binding site-modulating activity protein; EMA; MGC111545; Transcription factor E2F6; transcriptional repressor E2F6
Gene Symbols E2F6
Host Species Rabbit
Immunogen Recombinant Protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1876
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.