missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E2F7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114731
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total E2F7.
E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression.
Specifications
| E2F7 | |
| Polyclonal | |
| Unconjugated | |
| E2F7 | |
| A630014C11Rik; D10Ertd739e; E2F transcription factor 7; E2f7; E2F-7; fc43h03; FLJ12981; no; si:dkey-217k21.2; Transcription factor E2F7; wu:fc43h03 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 144455, 314818, 52679 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| D4A4D7, Q6S7F2, Q96AV8 | |
| E2F7 | |
| A synthesized peptide derived from human E2F7(Accession Q96AV8), corresponding to amino acid residues N33-P83. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction