missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F8 Polyclonal Antibody

E2F8 Polyclonal Antibody
Brand: Invitrogen PA566732
This item is not returnable.
View return policy
Description
E2F8 Polyclonal Antibody for ICC/IF
Specifications
| E2F8 | |
| Polyclonal | |
| Unconjugated | |
| E2F8 | |
| 4432406C08Rik; AA410048; E2F 8; E2F family member 8; E2F transcription factor 8; E2F8; E2F-8; FLJ23311; RGD1308091; Transcription factor E2F8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A0AVK6 | |
| E2F8 | |
| Recombinant Human E2F8. Recombinant protein control fragment (Product #RP-107384). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction