missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E6AP/UBE3A Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98953APC
This item is not returnable.
View return policy
Description
E6AP/UBE3A Polyclonal antibody specifically detects E6AP/UBE3A in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Immunohistochemistry (Paraffin) | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction