missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E6AP/UBE3A Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-239JF549
This item is not returnable.
View return policy
Description
E6AP/UBE3A Polyclonal antibody specifically detects E6AP/UBE3A in Human, Mouse samples. It is validated for Western Blot
Specifications
| Immunogen affinity purified | |
| RUO | |
| Purified |
| 0.1 mL | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction