missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E6AP/UBE3A Antibody (PCRP-UBE3A-1A2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08913UV
This item is not returnable.
View return policy
Description
E6AP/UBE3A Monoclonal antibody specifically detects E6AP/UBE3A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| E6AP/UBE3A | |
| Monoclonal | |
| DyLight 350 | |
| ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A | |
| Recombinant full-length human E6AP/UBE3A protein (Uniprot: Q05086) | |
| 100 μg | |
| Apoptosis, Transcription Factors and Regulators | |
| 7337 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-UBE3A-1A2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction