missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E6AP/UBE3A Antibody (SR1023), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 449.00
Specifications
| Antigen | E6AP/UBE3A |
|---|---|
| Clone | SR1023 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627524
|
Novus Biologicals
NBP3-21827-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600214
|
Novus Biologicals
NBP3-21827-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
E6AP/UBE3A Monoclonal antibody specifically detects E6AP/UBE3A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| E6AP/UBE3A | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A | |
| A synthesized peptide derived from human E6AP/UBE3A (Uniprot #: Q05086) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1023 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 7337 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title