missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E7 Antibody (6F3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37628
This item is not returnable.
View return policy
Description
E7 Monoclonal specifically detects E7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| E7 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| Mouse | |
| 21 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Ascites |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6F3 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| E7 transforming protein | |
| Purified recombinant fragment of E7 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 1489079 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction