missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EAAC1 Monoclonal Antibody (N180/41), PerCP

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545549
This item is not returnable.
View return policy
Description
5 μg of MA5-45549 was sufficient for immunoprecipitation of EAAC1 from 50 μg of rat brain lysate. Detects approximately 60kDa. This antibody was formerly sold as clone S180-41.
L-glutamate is the major excitatory neurotransmitter in mammalian central nervous systems. To maintain glutamate levels below excitoxic levels, excess glutamate at excitatory synaptic clefts is removed by ion-coupled glutamate transporters. Four glutamate transporters have been identified: the sodium-dependent GLAST-1, GLT-1, EAAC-1 and the chloride-dependent EAAT-4. EAAC-1 is expressed in the CNS and also found in kidney and small intestine. Structural features of glutamate transporters are believed to include eight membrane-spanning alpha-helices and a loop-pore structure which is unique among secondary transporters but may resemble loop-pores found in ion channels. A second distinctive structural feature is the presence of a highly amphipathic membrane-spanning helix that provides a hydrophilic path through the membrane. Mice with homozygous deletions of the EAAC-1 transporter gene develop dicarboxylic aminoaciduria and display reduced spontaneous locomotor activity although no neurodegeneration was observed over a period of 12 months.
Specifications
| EAAC1 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| P43005, P51907 | |
| Slc1a1 | |
| Fusion protein amino acids 1-524 (full length) of human EAAC1. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| N180/41 | |
| PerCP | |
| Slc1a1 | |
| D130048G10Rik; DCBXA; EAAC1; EAAC2; EAAT3; excitatory amino acid carrier 1; excitatory amino acid carrier 2; excitatory amino acid transporter 3; excitatory amino acid transporter-3; excitatory amino-acid carrier 1; glutamate transporter mEAAC2; MEAAC1; Neuronal and epithelial glutamate transporter; REAAC1; SCZD18; SLC1A1; Sodium-dependent glutamate/aspartate transporter 3; solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1; Solute carrier family 1 A1 (brain glutamate transporter); solute carrier family 1 member 1; solute carrier family 1, member 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 25550, 6505 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction