missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT1/GLAST-1/SLC1A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98747-50ul
This item is not returnable.
View return policy
Description
EAAT1/GLAST-1/SLC1A3 Polyclonal antibody specifically detects EAAT1/GLAST-1/SLC1A3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EAAT1/GLAST-1/SLC1A3 | |
| Polyclonal | |
| Immunohistochemistry 1:1000-1:4000, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLASTGLAST1, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the MOUSE EAAT1/GLAST-1/SLC1A3. | |
| 50 μg | |
| Cellular Markers, Cognition and Behavior | |
| 6507 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction