missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT2/GLT1 Antibody (SR2082), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21735-50ul
This item is not returnable.
View return policy
Description
EAAT2/GLT1 Monoclonal antibody specifically detects EAAT2/GLT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| EAAT2/GLT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR2082 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
| A synthesized peptide derived from human EAAT2/GLT1 (Uniprot #: P43004) | |
| 50 μg | |
| Hypoxia, Neuroscience, Neurotransmission | |
| 6506 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction