missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT3 Antibody (4D6.2), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29747IR
This item is not returnable.
View return policy
Description
EAAT3 Monoclonal antibody specifically detects EAAT3 in Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| EAAT3 | |
| Monoclonal | |
| DyLight 755 | |
| EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1 | |
| C-terminus peptide (KDKSDTISFTQTSQF). | |
| 0.1 mL | |
| ABC Transporters, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
| 6505 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 4D6.2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction