missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EAF1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16666974
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16666974 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16666974 Supplier Invitrogen™ Supplier No. PA5100493

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EAF1.

Acts as a transcriptional transactivator of ELL and ELL2 elongation activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAF1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EAF1
Gene Accession No. Q96JC9, Q9D4C5
Gene Alias 4933403C17Rik; AI666536; EAF1; ELL (eleven nineteen lysine-rich leukemia gene)-associated factor 1; ELL associated factor 1; ELL-associated factor 1
Gene Symbols EAF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human EAF1(Accession Q96JC9), corresponding to amino acid residues T133-I183.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306261, 74427, 85403
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.