missing translation for 'onlineSavingsMsg'
Learn More

EAF2 Antibody, Novus Biologicals™

Product Code. 18200676 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18200676 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18200676 Supplier Novus Biologicals Supplier No. NBP180575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EAF2 Polyclonal specifically detects EAF2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_060926.2
Gene Alias BM040, ELL associated factor 2, testosterone regulated apoptosis inducer and tumor suppressor, Testosterone-regulated apoptosis inducer and tumor suppressor protein, TRAITSELL-associated factor 2, U19
Gene Symbols EAF2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 150 and 200 of human ELL associated factor 2 using the numbering given in entry NP_060926.2 (GeneID 55840).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55840
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.