missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAF6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26465-1-AP-20UL
This item is not returnable.
View return policy
Description
The screening of cDNA expression libraries from human tumors with serum antibody (SEREX) has proven to be a powerful method for identifying the repertoire of tumor antigens recognized by the immune system of cancer patients, referred to as the cancer immunome. In this regard, cancer/testis (CT) antigens are of particular interest because of their immunogenicity and restricted expression patterns. Synovial sarcomas are striking with regard to CT antigen expression, however, highly expressed in sarcoma, CT antigens do not induce frequent humoral immune responses in sarcoma patients. Sera from two patients were used to immunoscreen cDNA libraries from two synovial sarcoma cell lines and normal testis, resulting in the identification of 113 distinct antigens.Specifications
| EAF6 | |
| Polyclonal | |
| Unconjugated | |
| MEAF6 | |
| C1orf149, CDABP0189, Eaf6, hEAF6, MEAF6, MYST/Esa1 associated factor 6, NY SAR 91, Protein EAF6 homolog, Sarcoma antigen NY SAR 91 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64769 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9HAF1 | |
| MEAF6 | |
| EAF6 Fusion Protein Ag23971 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction