missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EAPP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144994
This item is not returnable.
View return policy
Description
Immunogen sequence: KDNRDQAWVD AQRRGYHGLG PQRSRQQQPV PNSDAVLNCP ACMTTLCLDC QRHESYKTQY RAMFVMNCSI NK Antigen sequence identity: Mouse 90%; Rat 88%.
This protein may play an important role in the fine-tuning of both major E2F1 activities, the regulation of the cell-cycle and the induction of apoptosis. Promotes S-phase entry, and inhibits p14(ARP) expression. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. The C-terminal of EAPP binds the N-terminal of E2F1 and also interacts with E2F2 and E2F3, but not E2F4.
Specifications
| EAPP | |
| Polyclonal | |
| Unconjugated | |
| EAPP | |
| 1810011O16Rik; BM036; BM-036; c14orf11; E2F associated phosphoprotein; E2F-associated phosphoprotein; Eapp; hypothetical protein LOC327318; RGD1309624; Unknown (protein for MGC:139672); wu:fd21g02; zgc:65820 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55837 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q56P03 | |
| EAPP | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 143-214. Recombinant protein control fragment (Product #RP-109806). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction