Learn More
EARS2 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810468
Encompass_Preferred
 View more versions of this product
Product Code. 17543574
  • € 540.87 / 100µL
Description
Glutamyl-tRNA synthetase (GluRS or EARS2) a class I aminoacyl-tRNA synthetase (aaRS), is primarily responsible for the glutamylation of tRNAGlu. It is part of the ""minimal set"" of seventeen aaRSs found in every living organism and its presence is essential for the viability of cells.
Specifications
Specifications
EARS2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q5JPH6
EARS2
Full length human recombit protein of human EARS2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2B1
Unconjugated
EARS2
COXPD12, MSE1, gluRS
Mouse
Affinity Chromatography
RUO
124454
-20° C, Avoid Freeze/Thaw Cycles