Learn More
Abeba™ Easy 37630 Shoes
Product Code. 15671128
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15671128
Brand: Abeba™ 37630/46
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather, perforated
  • White
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color White
Material Leather
Size 46
Sole Features Slip-resistant
Sole Material Polyurethane
Style 37630