missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One EasyLoad™ 200 Refill System

Product Code. 10227161
Click to view available options
Color:
Natural
Sterility:
Non-sterile
Unit Size:
Pack of 5760
Pack of 960
This item is not returnable. View return policy

Product Code. 10227161

Brand: Greiner Bio-One 741000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Refill system for standard pipette tips, Universal 200μL, thin walled tips with graduation

  • PP (Polypropylene) tips
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Sterility Non-sterile
Color Natural
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line EasyLoad
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.