missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human APC Positive Selection Kit II
Description
- The EasySep™ Human APC Positive Selection Kit II is designed to isolate cells that are labeled with APC (allophycocyanin)-conjugated antibodies by positive selection
- Desired cells are targeted with antibody complexes recognizing APC and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- This product replaces the EasySep™ Human APC Positive Selection Kit (Catalog #18451)
Specifications
Specifications
| Cell Type | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?