missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Mouse CD19 Positive Selection Kit II

Product Code. 16695135
Encompass
Change view
Click to view available options
Quantity:
2 x 109 cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16695135 2 x 109 cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16695135 Supplier STEMCELL Technologies Supplier No. 18954

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic positive selection of CD19+ cells (e.g. B cells) from mouse splenocytes or other tissues

  • The EasySep™ Mouse CD19 Positive Selection Kit II is designed to isolate CD19+ cells from single-cell suspensions of splenocytes or other tissues by positive selection
  • Desired cells are targeted with antibody complexes recognizing CD19 and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • This product replaces the EasySep™ Mouse CD19 Positive Selection Kit (Catalog #18754) for even faster cell isolations and does not result in the labeling of isolated cells with PE

Spezifikation

Cell Type B Cells
For Use With (Application) Cell Isolation
Quantity 2 x 109 cells
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.