missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Mouse TIL (CD45) Positive Selection Kit

Product Code. 17167571
Encompass
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17167571

Brand: STEMCELL Technologies 1000350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic positive selection of mouse CD45+ TILs from single-cell suspensions of solid tumors

  • The EasySep™ Mouse TIL (CD45) Positive Selection Kit is designed to isolate highly purified CD45+ tumor infiltrating leukocytes (TILs) by immunomagnetic positive selection
  • This kit has been optimized for use on single-cell suspensions of solid mouse tumors with a low CD45+ start cell frequency, including tumors induced by implantation of 4T1, B16-F10, and CT26
  • WT cell lines into syngeneic mice
  • Due to the heterogeneity of mouse tumors, this kit may require optimization
  • This kit targets CD45+ cells for positive selection with antibodies recognizing the CD45 surface marker
  • Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
  • Unwanted cells are simply poured off, while desired cells remain in the tube
  • Isolated cells are immediately available for downstream applications such as flow cytometry, culture, and cell-based experiments

Specifications

Cell Type Leukocytes
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.