Learn More
STEMCELL Technologies EasySep™ Release Human CD19 Positive Selection Kit
Product Code. 17197751
Encompass
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Product Code. 17197751
Brand: STEMCELL Technologies 17754
Immunomagnetic positive selection of particle-free human CD19+ cells
  • The EasySep™ Release Human CD19 Positive Selection Kit is designed to isolate CD19+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection
  • Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
  • Unwanted cells are simply poured off, while desired cells remain in the tube
  • Then, bound magnetic particles are removed from the EasySep™-isolated CD19+ cells, which are immediately available for downstream applications
  • Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands
  • The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells
  • It is also expressed on follicular dendritic cells
Specifications
Cell Type B Cells
Quantity 1 x 109 cells