missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EB1/EB2/EB3 Monoclonal Antibody (KT65)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA547713

Product Code. 17817088

  • € 484.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The antibody detects a 30 kDa* protein corresponding to the molecular mass of EB1 on SDS-PAGE immunoblots of human HeLa, mouse brain, and rat PC12 cells. The antibody also detects EB1 by immunocytochemistry in HeLa and A431 cells. The antibody may weakly detect endogenous EB2 and EB3 in some cells and tissues.

The microtubule (MT) plus-end is a crucial site for the regulation of MT dynamics and interactions by several groups of plus-end tracking proteins (+TIPs). These +TIPs form comet-like accumulations at the plus ends of MTs to regulate MT dynamics and interactions with organelles and macromolecular complexes. The +TIPs include diverse groups of proteins, such as motor and nonmotor proteins, MT polymerases and depolymerases as well as various regulatory and adaptor proteins. The CLIP-associated protein (CLASP) family includes CLASP1 and CLASP2 proteins, which are expressed as long (alpha) and short (beta) isoforms. Thse +TIPs conatin an N-terminal TOG domain, multiple TOG-like domains, and a basic and serine-rich motif (SxIP). The TOG domain facilitates interaction with tubulin dimers, while the SxIP motif promotes interaciton with EB1 and MTs. A C-terminal domain is involved in interaction with CLIPs, as well as several other proteins. CLASPs are MT stabilizing fators that localize to mitotic spindles, kinetochores, and the midbody. CLASPs are important for cell division, and may regulate cell migration and neuronal growth cone motility.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EB1/EB2/EB3
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
Q15555, Q15691, Q3B8Q0, Q5XIT1, Q61166, Q66HR2, Q6PER3, Q8R001, Q9UPY8
Mapre1, Mapre2, Mapre3
Clone KT65 was generated from a GST fusion protein containing amino acids from the C-terminal region mouse EB1. This sequence is highly conserved in human and rat EB1.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Western Blot, Immunocytochemistry
KT65
Unconjugated
Mapre1
APC; Microtubule RP/EB
Rat
Protein G
RUO
100732, 10982, 114764, 13589, 212307, 22919, 22924, 298848, 679221
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.